Invitrogen
Manufacturer Code:PA527498
Catalog # PIPA527498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 227 and 447 of Human Rab GDI alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1A GDIL MRX41 MRX48 OPHN2 RABGD1A RABGDIA XAP-4 GDI-1 guanosine diphosphate dissociation inhibitor 1 mental retardation X-linked 48 oligophrenin-2 protein XAP-4 rab GDI alpha rab GDP dissociation inhibitor alpha rab GDP-dissociation inhibitor alpha; GDI-1; Guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; Oligophrenin-2; Protein XAP-4; Rab GDI alpha; Rab GDIalpha; Rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; testis secretory sperm-binding protein Li 208a
Gene Aliases: 1A; GDI1; GDIA; GDIalpha; GDIL; MRX41; MRX48; OPHN2; RABGD1A; RABGDIA; XAP-4; XAP4
UniProt ID: (Human) P31150, (Mouse) P50396
Entrez Gene ID: (Human) 2664, (Mouse) 14567
Molecular Function: G-protein modulator acyltransferase enzyme modulator transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.