Proteintech
Manufacturer Code:191421AP150UL
Catalog # 50-173-2355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GDF8/Myostatin Fusion Protein Ag13605 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: compact; GDF 8 GDF8 MSTN myostatin; GDF-8; growth differentiation factor 8; Growth/differentiation factor 8; Myostatin
Gene Aliases: Cmpt; GDF8; MSLHP; MSTN
UniProt ID: (Human) O14793, (Mouse) O08689
Entrez Gene ID: (Human) 2660, (Mouse) 17700
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.