Abnova
Manufacturer Code:PAB8589
Catalog # 89-133-769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human GDF6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant GDF6.
For Research Use Only
Protein Aliases: BMP-13; BMP13; Bone morphogenetic protein 13; CDMP2; GDF-6; Growth/differentiation factor 16; Growth/differentiation factor 6; KFS; KFSL; Klip-Feil malformation; Klippel-Feil malformation; Klippel-Feil syndrome; MGC158100; MGC158101; SGM1
Gene Aliases: BMP-13; BMP13; CDMP2; GDF16; GDF6; KFM; KFS; KFS1; KFSL; LCA17; MCOP4; MCOPCB6; SCDO4; SGM1
UniProt ID: (Human) Q6KF10
Entrez Gene ID: (Human) 392255
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.