Invitrogen
Manufacturer Code:PA547974
Catalog # PIPA547974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse GDF-3 Ala253-Gly366 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GDF-3; Growth/differentiation factor 3; KFS3 MCOP7 MCOPCB6 GDF-3 growth/differentiation factor 3; VG-1-related protein 2
Gene Aliases: C78318; ecat9; Gdf-3; GDF3; KFS3; MCOP7; MCOPCB6; UNQ222/PRO248; Vgr-2; Vgr2
UniProt ID: (Human) Q9NR23, (Mouse) Q07104
Entrez Gene ID: (Human) 9573, (Mouse) 14562
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.