Invitrogen
Manufacturer Code:MA537793
Catalog # PIMA537793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2182CT228.18.8 |
Immunogen | recombinant protein from the human GDF15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GDF-15; GDF15 GDF-15 growth differentiation factor 15 growth/differentiation factor 15 macrophage inhibiting cytokine-1 macrophage inhibitory cytokine 1 MIC1 MIC-1 NAG1 NAG-1 NRG-1 NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1 NSAID-activated gene 1 protein NSAID-regulated gene 1 protein PDF PLAB Placental bone morphogenetic protein placental TGF-beta Prostate differentiation factor PTGFB PTGF-beta Sbf; Growth/differentiation factor 15; Macrophage inhibitory cytokine 1; MIC-1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; Placental bone morphogenetic protein; Placental TGF-beta; Prostate differentiation factor; PTGF-beta
Gene Aliases: GDF-15; GDF15; MIC-1; MIC1; NAG-1; PDF; PLAB; PTGFB
UniProt ID: (Human) Q99988
Entrez Gene ID: (Human) 9518
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.