Novus Biologicals
Manufacturer Code:NBP212334AF700
Catalog # NB212334700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0243-13F2 |
Immunogen | Mouse GDF8/Myostatin |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: GDF-8; GDF-8 GDF8growth differentiation factor 8 growth/differentiation factor 8 Myostatin; Growth/differentiation factor 8; Myostatin
Gene Aliases: GDF8; MSLHP; MSTN
UniProt ID: (Human) O14793
Entrez Gene ID: (Human) 2660
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.