R&D Systems
Manufacturer Code:AF1539SP
Catalog # AF1539SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse GDF-8/Myostatin Propeptide Asn25-Ser265 Accession # O08689 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: GDF-8; GDF8 GDF-8 GDF8growth differentiation factor 8 growth/differentiation factor 8 MSLHP MSTN Myostatin; Growth/differentiation factor 8; Myostatin
Gene Aliases: GDF8; MSLHP; MSTN
UniProt ID: (Human) O14793
Entrez Gene ID: (Human) 2660
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.