Novus Biologicals
Manufacturer Code:NBP176364
Catalog # NBP176364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human GDF6. The immunogen is located within amino acids 320 - 370 of GDF6. Amino Acid Squence: AFASRHGKRHGKKSRLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-13; BMP13 CDMP2 GDF16 GDF-6 growth differentiation factor 6 Growth/differentiation factor 16 growth/differentiation factor 6 KFS KFS1 KFSL MCOP4 MCOPCB6 MGC158100 MGC158101 SCDO4 SGM1; Bone morphogenetic protein 13; GDF-6; Growth/differentiation factor 16; Growth/differentiation factor 6; Klip-Feil malformation; Klippel-Feil malformation; Klippel-Feil syndrome
Gene Aliases: BMP-13; BMP13; CDMP2; GDF16; GDF6; KFM; KFS; KFS1; KFSL; LCA17; MCOP4; MCOPCB6; SCDO4; SGM1
UniProt ID: (Human) Q6KF10
Entrez Gene ID: (Human) 392255
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.