Novus Biologicals
Manufacturer Code:NBP212333V
Catalog # NBP212333V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0242-13J15 |
Immunogen | Mouse GDF5 |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: BMP-14; BMP14 Cartilage-derived morphogenetic protein 1 cartilage-derived morphogenetic protein-1 CDMP-1 CDMP1LAP4 GDF-5 growth differentiation factor 5 growth/differentiation factor 5 OS5 Radotermin SYNS2; Bone morphogenetic protein 14; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP-1; GDF-5; Growth/differentiation factor 5; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; Radotermin
Gene Aliases: BDA1C; BMP-14; BMP14; CDMP1; GDF5; LAP-4; LAP4; OS5; SYM1B; SYNS2
UniProt ID: (Human) P43026
Entrez Gene ID: (Human) 8200
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.