Novus Biologicals
Manufacturer Code:NBP299527R
Catalog # NB061348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse GDF-3. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDF-3; GDF-3 growth differentiation factor 3 growth/differentiation factor 3 KFS3 MCOP7 MCOPCB6; Growth/differentiation factor 3
Gene Aliases: GDF3; KFS3; MCOP7; MCOPCB6; UNQ222/PRO248
UniProt ID: (Human) Q9NR23
Entrez Gene ID: (Human) 9573
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.