Novus Biologicals
Manufacturer Code:NBP212331R
Catalog # NBP212331R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0307-10A3 |
Immunogen | Human GDF15 |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GDF-15; growth differentiation factor 15 growth/differentiation factor 15 Macrophage inhibitory cytokine 1 MIC-1NSAID-activated gene 1 protein MIC1Prostate differentiation factor NAG-1NSAID-regulated gene 1 protein NSAID (nonsteroidal inflammatory drug)-activated protein 1 PDFGDF-15 PLABNRG-1 Placental bone morphogenetic protein Placental TGF-beta PTGFBPTGF-beta; Growth/differentiation factor 15; Macrophage inhibitory cytokine 1; MIC-1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; Placental bone morphogenetic protein; Placental TGF-beta; Prostate differentiation factor; PTGF-beta
Gene Aliases: GDF-15; GDF15; MIC-1; MIC1; NAG-1; PDF; PLAB; PTGFB
UniProt ID: (Human) Q99988
Entrez Gene ID: (Human) 9518
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.