R&D Systems
Manufacturer Code:AF957SP
Catalog # AF957SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human GDF-15 Ala197-Ile308 Accession # Q99988 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GDF-15; GDF15 growth differentiation factor 15 growth/differentiation factor 15 Macrophage inhibitory cytokine 1 MIC-1 MIC-1NSAID-activated gene 1 protein MIC1Prostate differentiation factor NAG-1 NAG-1NSAID-regulated gene 1 protein NSAID (nonsteroidal inflammatory drug)-activated protein 1 PDF PDFGDF-15 PLAB PLABNRG-1 Placental bone morphogenetic protein Placental TGF-beta PTGF-beta PTGFBPTGF-beta; Growth/differentiation factor 15; Macrophage inhibitory cytokine 1; MIC-1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; Placental bone morphogenetic protein; Placental TGF-beta; Prostate differentiation factor; PTGF-beta
Gene Aliases: GDF-15; GDF15; MIC-1; MIC1; NAG-1; PDF; PLAB; PTGFB
UniProt ID: (Human) Q99988
Entrez Gene ID: (Human) 9518
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.