Proteintech
Manufacturer Code:112722AP150UL
Catalog # 50-172-7019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GDE1 Fusion Protein Ag1821 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 363E6.2 GDE1 MIR16; Glycerophosphodiester phosphodiesterase 1; Glycerophosphoinositol glycerophosphodiesterase GDE1; Lysophospholipase D GDE1; membrane interacting protein of RGS16; Membrane-interacting protein of RGS16; RGS16-interacting membrane protein
Gene Aliases: 1200003M13Rik; 363E6.2; GDE1; MIR16; RGS16
UniProt ID: (Human) Q9NZC3, (Rat) Q9JL55, (Mouse) Q9JL56
Entrez Gene ID: (Human) 51573, (Rat) 60418, (Mouse) 56209
Molecular Function:
esterase
hydrolase
ligase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.