Invitrogen
Manufacturer Code:PA579304
Catalog # PIPA579304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human GDA recombit protein (Position: E220-Q446). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAH; Guanase; guanine aminase; Guanine aminohydrolase; Guanine deaminase; Guanine deaminase Guanase Guanine aminase Guanine aminohydrolase GAH p51-nedasin GDA KIAA1258
Gene Aliases: AU015411; AW047581; Cypin; Gda
UniProt ID: (Mouse) Q9R111
Entrez Gene ID: (Mouse) 14544, (Rat) 83585
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.