Invitrogen
Manufacturer Code:PA526715
Catalog # PIPA526715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 94-122 amino acids from the N-terminal region of human GDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYPIN GUANASE NEDASIN GAH cytoplasmic PSD95 interactor guanine aminase guanine aminohydrolase p51-nedasin; cytoplasmic PSD95 interactor; GAH; Guanase; guanine aminase; Guanine aminohydrolase; Guanine deaminase; p51-nedasin
Gene Aliases: CYPIN; GDA; GUANASE; KIAA1258; NEDASIN
UniProt ID: (Human) Q9Y2T3
Entrez Gene ID: (Human) 9615
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.