Novus Biologicals
Manufacturer Code:NBP290289AF405
Catalog # NB071889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 105 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human GCSH (AAA36011.1 Ser49-Glu173). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: GCE glycine cleavage system H protein mitochondrial glycine cleavage system protein H (aminomethyl carrier) lipoic acid-containing protein mitochondrial glycine cleavage system H-protein NKH; Glycine cleavage system H protein, mitochondrial; glycine cleavage system protein H (aminomethyl carrier); Lipoic acid-containing protein; mitochondrial glycine cleavage system H-protein
Gene Aliases: GCE; GCSH; NKH
UniProt ID: (Human) P23434
Entrez Gene ID: (Human) 2653
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.