Invitrogen
Manufacturer Code:PA568108
Catalog # PIPA568108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human GCNT4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2GNT3 beta-13-galactosyl-O-glycosyl-glycoprotein beta-16-N-acetylglucosaminyltransferase 4 core 2 beta-16-N-acetylglucosaminyltransferase 3 core 2-branching enzyme 3 core2-GlcNAc-transferase 3 glucosaminyl (N-acetyl) transferase 4 core 2 (beta-16-N-acetylg; core 2 beta-1,6-N-acetylglucosaminyltransferase 3; core 2-branching enzyme 3; core2-GlcNAc-transferase 3; glucosaminyl (N-acetyl) transferase 4, core 2 (beta-1,6-N-acetylglucosaminyltransferase)
Gene Aliases: C2GNT3
Entrez Gene ID: (Human) 51301
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.