Invitrogen
Manufacturer Code:PA524455
Catalog # PIPA524455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 390-418 amino acids from the C-terminal region of human GCNT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3; beta1,6-N-acetylglucosaminyltransferase-M; beta1,6GlcNAc-transferase; C2/4GnT; C2/4GnT C24GNT C2GNT2 C2GNTM GNTM C2GnT-mucin type beta-13-galactosyl-O-glycosyl-glycoprotein beta-16-N-acetylglucosaminyltransferase 3 beta16-N-acetylglucosaminyltransferase-M beta16GlcNAc-transferase core 2/core 4 beta-16-N-acetylglucosaminyltransferase hC2GnT-M mucus-type core 2 b; C2GnT-M; C2GnT-mucin type; core 2 beta-1,6-N-acetylglucosaminyltransferase II; Core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; hC2GnT-M; Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase
Gene Aliases: 2010013H22Rik; 2210021I22Rik; 2210401J11Rik; C2/4GnT; C24GNT; C2GNT2; C2GNTM; GCNT3; GNTM
UniProt ID: (Human) O95395, (Mouse) Q5JCT0
Entrez Gene ID: (Human) 9245, (Mouse) 72077
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.