Novus Biologicals
Manufacturer Code:NBP188313
Catalog # NBP188313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RLKCDSDHLGLESRESQSQYCRNILYNFLKLPAKRSINCSGVTRGDQEAVLQAILNNLEVKKKREPFTDTHYLSLTRDCEHFKAERKFIQFPLSKEEVEFPIAYSMVIHEKIENFER |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3; beta-13-galactosyl-O-glycosyl-glycoproteinbeta-16-N-acetylglucosaminyltransferase 3 beta16GlcNAc-transferase beta16-N-acetylglucosaminyltransferase-M C2/4GnTGNTM C24GNT C2GnT2 C2GnT-M C2GNTM C2GnT-mucin type Core 2/core 4 beta-16-N-acetylglucosaminyltransferase EC 2.4.1.102 EC 2.4.1.150 glucosaminyl (N-acetyl) transferase 3 mucin type hC2GnT-M mucus-type core 2 beta-16-N-acetylglucosaminyltransferase; beta1,6-N-acetylglucosaminyltransferase-M; beta1,6GlcNAc-transferase; C2/4GnT; C2GnT-M; C2GnT-mucin type; Core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; hC2GnT-M; mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase
Gene Aliases: C2/4GnT; C24GNT; C2GNT2; C2GNTM; GCNT3; GNTM
UniProt ID: (Human) O95395
Entrez Gene ID: (Human) 9245
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.