Novus Biologicals
Manufacturer Code:NBP255277
Catalog # NBP255277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QEFEEEANHWSNHGDSDHSSPGNRASESNQPSPGSTLSSSRSVELNGFMAFREQYMGMSVPPHYQYIPHLFSYSGHSPLLPQQARSLDPQSYSLIHQLLSAEDLEPLGTPMLIEDGYA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCNF; GCNF1 GCNFNR61 Germ cell nuclear factor hGCNF nuclear receptor subfamily 6 group A member 1 nuclear receptor subfamily 6 group A member 1 retinoic acid receptor-related testis-associated receptor Retinoid receptor-related testis-specific receptor RTRhRTR; Germ cell nuclear factor; germ cell nuclear factor variant 1; Nuclear receptor subfamily 6 group A member 1; nuclear receptor subfamily 6, group A, member 1; retinoic acid receptor-related testis-associated receptor; Retinoid receptor-related testis-specific receptor; RTR
Gene Aliases: CT150; GCNF; GCNF1; hGCNF; hRTR; NR61; NR6A1; RTR
UniProt ID: (Human) Q15406
Entrez Gene ID: (Human) 2649
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.