Invitrogen
Manufacturer Code:MA514884
Catalog # PIMA514884
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | S.775.4 |
Immunogen | Synthetic peptide corresponding to the amino terminus of human GCN5L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: GCN5 (general control of amino-acid synthesis yeast homolog)-like 2 General control of amino acid synthesis yeast homolog-like 2 STAF97 general control of amino acid synthesis protein 5-like 2 histone acetyltransferase GCN5 histone acetyltransferase KAT2A hsGCN5 lysine acetyltransferase 2A; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2; GCN5 general control of amino acid synthesis-like 2; general control of amino acid synthesis 5-like 2; General control of amino acid synthesis protein 5-like 2; general control of amino acid synthesis, yeast homolog-like 2; General control of amino acid synthesis, yeast, homolog-like 2; general control of amino acid synthesis-like 2; hGCN5; Histone acetyltransferase GCN5; Histone acetyltransferase KAT2A; Histone glutaryltransferase KAT2A; Histone succinyltransferase KAT2A; hsGCN5; K(lysine) acetyltransferase 2A; Lysine acetyltransferase 2A; MmGCN5; PCAF-B/GCN5; STAF97
Gene Aliases: 1110051E14Rik; AW212720; GCN5; GCN5L2; hGCN5; KAT2A; mmGCN5; PCAF-b
UniProt ID: (Human) Q92830, (Mouse) Q9JHD2
Entrez Gene ID: (Human) 2648, (Horse) 100053057, (Mouse) 14534, (Rat) 303539
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.