Abnova
Manufacturer Code:PAB1162
Catalog # 89-115-226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 1-300 <i>Saccharomyces cerevisiae</i> GCN5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae GCN5.
For Research Use Only
Protein Aliases: ADA4; Histone acetyltransferase GCN5; SWI9
Gene Aliases: AAS104; ADA4; GCN5; KAT2; SWI9; YGR252W
UniProt ID: (Baker's yeast) Q03330
Entrez Gene ID: (Baker's yeast) 853167
Molecular Function: DNA binding protein acetyltransferase chromatin/chromatin-binding protein nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.