Novus Biologicals
Manufacturer Code:NBP281273SS
Catalog # NB016853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C11L34 |
Immunogen | Biotinylated GCN4-peptide (HLENEVARLKK). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for up to 3 months. For longer storage aliquot and store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AAS101 AAS3 Amino acid biosynthesis regulatory protein amino acid starvation-responsive transcription factor GCN4 ARG9 General Control Protein 4; Amino acid biosynthesis regulatory protein; amino acid starvation-responsive transcription factor GCN4; General control protein GCN4; General control transcription factor GCN4
Gene Aliases: AAS101; AAS3; ARG9; GCN4; YEL009C
UniProt ID: (Baker's yeast) P03069
Entrez Gene ID: (Baker's yeast) 856709
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.