R&D Systems
Manufacturer Code:MAB6878
Catalog # MAB6878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 708210 |
Immunogen | <EM>E. coli</EM>-derived recombinant human GCN2 Arg22-Lys139 Accession # Q9P2K8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-alpha kinase GCN2; EIF2AK4 eukaryotic translation initiation factor 2 alpha kinase 4 eukaryotic translation initiation factor 2-alpha kinase 4 GCN2GCN2 eIF2alpha kinase GCN2-like protein KIAA1338EC 2.7.11.1; Eukaryotic translation initiation factor 2-alpha kinase 4; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2
Gene Aliases: EIF2AK4; GCN2; KIAA1338; PVOD2
UniProt ID: (Human) Q9P2K8
Entrez Gene ID: (Human) 440275
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.