Invitrogen
Manufacturer Code:PA571908
Catalog # PIPA571908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1477-1504 amino acids from the C-terminal region of mouse Eif2ak4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-alpha kinase GCN2; Eukaryotic translation initiation factor 2-alpha kinase 4; Eukaryotic translation initiation factor 2-alpha kinase 4 GCN2-like protein mGCN2 Eif2ak4 Gcn2 Kiaa1338; GCN2 eIF2alpha kinase; GCN2-like protein; mGCN2
Gene Aliases: 2610011M03; Eif2ak4; GCN2; Kiaa1338; MGCN2
UniProt ID: (Mouse) Q9QZ05
Entrez Gene ID: (Mouse) 27103
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.