Invitrogen
Manufacturer Code:PA517523
Catalog # PIPA517523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to sequence near the amino terminus of human GCN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-alpha kinase GCN2; Eukaryotic translation initiation factor 2-alpha kinase 4; GCN2 eIF2alpha kinase; GCN2 eIF2alpha kinase GCN2-like protein eukaryotic translation initiation factor 2-alpha kinase 4; GCN2-like protein; general control nonderepressible 2; mGCN2
Gene Aliases: 2610011M03; EIF2AK4; GCN2; KIAA1338; MGCN2; PVOD2
UniProt ID: (Human) Q9P2K8, (Mouse) Q9QZ05
Entrez Gene ID: (Human) 440275, (Mouse) 27103, (Rat) 114859
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.