Invitrogen
Manufacturer Code:PA513924
Catalog # PIPA513924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GCN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-alpha kinase GCN2; Eukaryotic translation initiation factor 2-alpha kinase 4; Eukaryotic translation initiation factor 2-alpha kinase 4 GCN2-like protein EIF2AK4 GCN2 KIAA1338; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2
Gene Aliases: EIF2AK4; GCN2; KIAA1338; PVOD2
UniProt ID: (Human) Q9P2K8
Entrez Gene ID: (Human) 440275
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.