Novus Biologicals
Manufacturer Code:NBP183383
Catalog # NBP183383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EALVTDAGEVTEAGKAYVPPRVLQEALCVISGVPGLKGDVTDTEQLAQEMLIISHHPSLVAVQSGLWPALLARMKIDPEAFITRHLDQIIPRMTTQSPLNQSSMNAMGSLSVLSPDRVLPQLISTITA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-alpha kinase activator GCN1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF-2-alpha kinase activator homolog; GCN1 eIF2 alpha kinase activator homolog; GCN1 GCN1 (general control of amino-acid synthesis 1 yeast)-like 1 GCN1 general control of amino-acid synthesis 1-like 1 (yeast) GCN1L GCN1-like protein 1 HsGCN1 KIAA0219hsGCN1 peroxisome proliferator activated receptor interacting complex protein PRIC295 translational activator GCN1; GCN1 general control of amino-acid synthesis 1-like 1; GCN1-like protein 1; General control of amino-acid synthesis 1-like protein 1; HsGCN1; peroxisome proliferator activated receptor interacting complex protein; Translational activator GCN1
Gene Aliases: GCN1; GCN1L; GCN1L1; KIAA0219; PRIC295
UniProt ID: (Human) Q92616
Entrez Gene ID: (Human) 10985
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.