Abnova
Manufacturer Code:PAB21686
Catalog # 89-123-383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human GCN1L1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant GCN1L1.
For Research Use Only
Protein Aliases: eIF-2-alpha kinase activator GCN1; GCN1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF-2-alpha kinase activator homolog; GCN1 eIF2 alpha kinase activator homolog; GCN1 general control of amino-acid synthesis 1-like 1; GCN1-like protein 1; GCN1L; General control of amino-acid synthesis 1-like protein 1; HsGCN1; KIAA0219; peroxisome proliferator activated receptor interacting complex protein; Translational activator GCN1
Gene Aliases: GCN1; GCN1L; GCN1L1; KIAA0219; PRIC295
UniProt ID: (Human) Q92616
Entrez Gene ID: (Human) 10985
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.