Invitrogen
Manufacturer Code:MA537836
Catalog # PIMA537836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0597 |
Immunogen | A synthesized peptide derived from human GCLM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI649393 gamma glutamylcysteine synthetase Gamma-ECS regulatory subunit gamma-glutamylcysteine synthetase regulatory subunit Gclm Gcmc GCS light chain GLCLR glutamate cysteine ligase (gamma-glutamylcysteine synthetase) regulatory glutamate cysteine ligase modifier subunit glutamate-cysteine ligase (gamma-glutamylcysteine synthetase) regulatory (30.8kD) glutamate-cysteine ligase modifier subunit glutamate--cysteine ligase modifier subunit glutamate-cysteine ligase modifier subunit delta2 alternative splicing glutamate-cysteine ligase regulatory protein Glutamate--cysteine ligase regulatory subunit glutamate-cysteine ligase modifier subunit GSC light chain null; Gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; GCS light chain; Glutamate--cysteine ligase modifier subunit; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; glutamate-cysteine ligase, modifier subunit; GSC light chain
Gene Aliases: GCLM; GLCLR
UniProt ID: (Human) P48507
Entrez Gene ID: (Human) 2730
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.