Invitrogen
Manufacturer Code:PA586516
Catalog # PIPA586516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 50-100 of Human GCSm-gamma. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; GCS light chain; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; Glutamate--cysteine ligase modifier subunit; Glutamate--cysteine ligase regulatory subunit; Glutamate--cysteine ligase regulatory subunit GCS light chain Gamma-ECS regulatory subunit Gamma-glutamylcysteine synthetase regulatory subunit Glutamate--cysteine ligase modifier subunit GLCLR GCLM; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; glutamate-cysteine ligase, modifier subunit; GSC light chain; null
Gene Aliases: AI649393; GCLM; Gcmc; GLCLR
UniProt ID: (Human) P48507, (Mouse) O09172, (Rat) P48508
Entrez Gene ID: (Human) 2730, (Mouse) 14630, (Rat) 29739
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.