Invitrogen
Manufacturer Code:PA526111
Catalog # PIPA526111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 246-274 amino acids from the C-terminal region of human GCLM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; GCS light chain; GLCLR GCS light chain GSC light chain gamma-ECS regulatory subunit gamma-glutamylcysteine synthetase regulatory subunit glutamate--cysteine ligase modifier subunit glutamate--cysteine ligase regulatory subunit glutamate-cysteine ligase (gamma-glutamylcysteine s; Glutamate--cysteine ligase modifier subunit; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; glutamate-cysteine ligase, modifier subunit; GSC light chain
Gene Aliases: AI649393; GCLM; Gcmc; GLCLR
UniProt ID: (Human) P48507, (Mouse) O09172
Entrez Gene ID: (Human) 2730, (Mouse) 14630
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.