Novus Biologicals
Manufacturer Code:NBP216624
Catalog # NBP216624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human GCLM. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gamma-ECS regulatory subunit; Gamma-ECS regulatory subunit Gamma-glutamylcysteine synthetase regulatory subunit GCS light chain GLCLRglutamate--cysteine ligase regulatory subunit glutamate-cysteine ligase (gamma-glutamylcysteine synthetase) regulatory(30.8kD) Glutamate--cysteine ligase modifier subunit glutamate-cysteine ligase regulatory protein glutamate-cysteine ligase modifier subunit GSC light chain; Gamma-glutamylcysteine synthetase regulatory subunit; GCS light chain; Glutamate--cysteine ligase modifier subunit; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; glutamate-cysteine ligase, modifier subunit; GSC light chain
Gene Aliases: GCLM; GLCLR
UniProt ID: (Human) P48507
Entrez Gene ID: (Human) 2730
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.