Invitrogen
Manufacturer Code:PA544190
Catalog # PIPA544190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GCLC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma GCS-HS; Gamma-ECS; Gamma-glutamylcysteine synthetase; gamma-glutamylcysteine synthetase heavy subunit; GCS heavy chain; Glutamate--cysteine ligase catalytic subunit; Glutamate-cysteine ligase catalytic subunit GCL GCS GLCL GLCLC; glutamate-cysteine ligase, catalytic subunit; Glutamylcysteine gamma synthetase light chain
Gene Aliases: cb1049; D9Wsu168e; fe36e11; GCL; GCLC; GCS; Ggcs-hs; GLCL; GLCL-H; GLCLC; wu:fe36e11
UniProt ID: (Human) P48506, (Rat) P19468, (Mouse) P97494
Entrez Gene ID: (Horse) 100056731, (Human) 2729, (Rabbit) 100342383, (Dog) 609822, (Bovine) 512468, (Rat) 25283, (Mouse) 14629, (Zebrafish) 326857
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.