Novus Biologicals
Manufacturer Code:NB600544
Catalog # NB600544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from corresponding to aa 295-313 of rat GCLC (heavy subunit of GCS) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.3.2.2 gamma-ECS Gamma-glutamylcysteine synthetase GCS GCS heavy chain GLCL GLCLCGCL glutamate--cysteine ligase catalytic subunit glutamate-cysteine ligase catalytic subunit; Gamma-ECS; Gamma-glutamylcysteine synthetase; GCS heavy chain; Glutamate--cysteine ligase catalytic subunit; glutamate-cysteine ligase, catalytic subunit
Gene Aliases: GCL; GCLC; GCS; GLCL; GLCLC
UniProt ID: (Human) P48506
Entrez Gene ID: (Human) 2729
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.