Invitrogen
Manufacturer Code:PA527959
Catalog # PIPA527959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 68 and 493 of Human GCKR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGQTL5 GKRP glucokinase regulatory protein; GKRP; glucokinase regulator; Glucokinase regulatory protein
Gene Aliases: FGQTL5; GCKR; GKRP
UniProt ID: (Human) Q14397, (Mouse) Q91X44
Entrez Gene ID: (Human) 2646, (Mouse) 231103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.