Invitrogen
Manufacturer Code:PA5109758
Catalog # PIPA5109758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GCKR (NP_001477.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGQTL5 GCKR GKRP GLRE Glucokinase regulator Glucokinase regulatory protein hexokinase 4; GKRP; Glucokinase regulator; Glucokinase regulatory protein
Gene Aliases: FGQTL5; GCKR; GKRP; GLRE
UniProt ID: (Human) Q14397, (Rat) Q07071, (Mouse) Q91X44
Entrez Gene ID: (Human) 2646, (Rat) 25658, (Mouse) 231103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.