OriGene
Manufacturer Code:CF500459
Catalog # 50-165-9531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3E3 |
Immunogen | Full length human recombit protein of human GCK produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP:D-hexose 6-phosphotransferase; FGQTL3 GK GLK HHF3 HK4 HKIV HXKP LGLK MODY2; Glucokinase; glucokinase (hexokinase 4); hexokinase D, pancreatic isozyme; Hexokinase type IV; Hexokinase-4; Hexokinase-D; HK IV; HK4
Gene Aliases: FGQTL3; GCK; GK; GLK; HHF3; HK4; HKIV; HXKP; LGLK; MODY2
UniProt ID: (Human) P35557
Entrez Gene ID: (Human) 2645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.