Proteintech
Manufacturer Code:196661AP150UL
Catalog # 50-173-2474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GCK Fusion Protein Ag8116 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCK GK GLK Glucokinase glucokinase (hexokinase 4) Hexokinase 4 Hexokinase D Hexokinase type IV HHF3 HK IV HK4 HKIV HXKP MODY2; Glucokinase; glucokinase variant 3; hexokinase 4; Hexokinase type IV; Hexokinase-4; Hexokinase-D; HK IV; HK4
Gene Aliases: Gck; Gk; GLK; Gls006; GLUKA; HK4; HKIV; Hlb62; HXKP; MODY2; RNGK2
UniProt ID: (Rat) P17712, (Mouse) P52792
Entrez Gene ID: (Rat) 24385, (Mouse) 103988
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.