Invitrogen
Manufacturer Code:PA5115273
Catalog # PIPA5115273
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GCHFR corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010323F13Rik GCHFR GFRP GTP cyclohydrolase 1 feedback regulatory protein GTP cyclohydrolase I feedback regulator GTP cyclohydrolase I feedback regulatory protein HsT16933 P35; GFRP; GTP cyclohydrolase 1 feedback regulatory protein; GTP cyclohydrolase I feedback regulatory protein; p35
Gene Aliases: 2010323F13Rik; GCHFR; GFRP; HsT16933; P35
UniProt ID: (Human) P30047, (Mouse) P99025, (Rat) P70552
Entrez Gene ID: (Human) 2644, (Mouse) 320415, (Rat) 171128
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.