OriGene
Manufacturer Code:CF810245
Catalog # 50-166-9966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A3 |
Immunogen | Full length human recombit protein of human GCH1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dystonia 14; DYT14 DYT5 DYT5a GCH GTP-CH-1 GTPCH1 HPABH4B; GTP cyclohydrolase 1; GTP cyclohydrolase I; GTP-CH-I; guanosine 5'-triphosphate cyclohydrolase I
Gene Aliases: DYT14; DYT5; DYT5a; GCH; GCH1; GTP-CH-1; GTPCH1; HPABH4B
UniProt ID: (Human) P30793
Entrez Gene ID: (Human) 2643
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.