Abnova
Manufacturer Code:H00002643M03
Catalog # 89-022-287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F5 |
Immunogen | GCH1 (AAH25415.1 1 a.a. ~ 250 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant GCH1.
For Research Use Only
Protein Aliases: dystonia 14; DYT14; DYT5; GCH; GTP cyclohydrolase 1; GTP cyclohydrolase I; GTP-CH-1; GTP-CH-I; GTPCH1; guanosine 5'-triphosphate cyclohydrolase I
Gene Aliases: DYT14; DYT5; DYT5a; GCH; GCH1; GTP-CH-1; GTPCH1; HPABH4B
UniProt ID: (Human) P30793
Entrez Gene ID: (Human) 2643
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.