Invitrogen
Manufacturer Code:PA569241
Catalog # PIPA569241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat GCDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD5 GCD glutaryl-CoA dehydrogenase mitochondrial glutaryl-Coenzyme A dehydrogenase; GCD; Glutaryl-CoA dehydrogenase, mitochondrial; glutaryl-Coenzyme A dehydrogenase
Gene Aliases: 9030411L18; ACAD5; AI266902; D17825; GCD; GCDH
UniProt ID: (Bovine) Q2KHZ9, (Human) Q92947, (Mouse) Q60759
Entrez Gene ID: (Dog) 476696, (Horse) 100063310, (Rabbit) 100343376, (Bovine) 506310, (Human) 2639, (Pig) 100522477, (Guinea pig) 100714939, (Mouse) 270076, (Rat) 364975
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.