Proteintech
Manufacturer Code:160681AP150UL
Catalog # 50-173-0689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GCDFP-15/PIP Fusion Protein Ag9004 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCDFP 15 GCDFP15 gp17 GPIP4 PIP prolactin induced protein Prolactin inducible protein SABP; GCDFP-15; gp17; Gross cystic disease fluid protein 15; Prolactin-induced protein; Prolactin-inducible protein; SABP; Secretory actin-binding protein
Gene Aliases: GCDFP-15; GCDFP15; GPIP4; PIP
UniProt ID: (Human) P12273
Entrez Gene ID: (Human) 5304
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.