Invitrogen
Manufacturer Code:PA590717
Catalog # PIPA590717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 170-419 of human GCAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-amino-3-ketobutyrate coenzyme A ligase, mitochondrial; 2-amino-3-ketobutyrate-CoA ligase; AKB ligase; Aminoacetone synthase; Glycine acetyltransferase; glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase)
Gene Aliases: AI526977; GCAT; KBL
UniProt ID: (Human) O75600, (Mouse) O88986
Entrez Gene ID: (Human) 23464, (Mouse) 26912, (Rat) 366959
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.