Novus Biologicals
Manufacturer Code:NBP181564
Catalog # NBP181564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:CLASRYGALVFMDECHATGFLGPTGRGTDELLGVMDQVTIINSTLGKALGGASGGYTTGPGPLVSLLRQRARPYLFSNSLPPAVVGCASKALDLLMGSNTIVQSMAAKTQRFRSKMEAAGFTISGA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-amino-3-ketobutyrate coenzyme A ligase, mitochondrial; 2-amino-3-ketobutyrate-CoA ligase; 2-amino-3-ketobutyrate-CoA ligase AKB ligase Aminoacetone synthase EC 2.3.1 EC 2.3.1.29 Glycine acetyltransferase glycine C-acetyltransferase glycine C-acetyltransferase (2-amino-3-ketobutyrate coenzyme A ligase) KBL2-amino-3-ketobutyrate coenzyme A ligase mitochondrial MGC23053; AKB ligase; Aminoacetone synthase; Glycine acetyltransferase
Gene Aliases: GCAT; KBL
UniProt ID: (Human) O75600
Entrez Gene ID: (Human) 23464
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.