Invitrogen
Manufacturer Code:PA545026
Catalog # PIPA545026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GUCA1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCAP 2; GCAP2 (retina); Guanylate cyclase activator 1B; guanylate cyclase activator 1B (retina); Guanylate cyclase activator 1B (retina) DKFZp686E1183 GCAP2 GUCA2 RP48; guanylate cyclase-activating protein, photoreceptor 2; Guanylyl cyclase-activating protein 2; Retinal guanylyl cyclase activator protein p24
Gene Aliases: BC018258; GCAP-2; GCAP2; GUCA1B; GUCA2; RP48
UniProt ID: (Bovine) P51177, (Human) Q9UMX6, (Mouse) Q8VBV8
Entrez Gene ID: (Bovine) 286851, (Rabbit) 100351008, (Horse) 100054522, (Dog) 481801, (Human) 2979, (Mouse) 107477, (Guinea pig) 100714867, (Rat) 316218
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.