Novus Biologicals
Manufacturer Code:NBP255158
Catalog # NBP255158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EYVAALSLVLKGKVEQKLRWYFKLYDVDGNGCIDRDELLTIIQAIRAINPCSDTTMTAEEFTDTVFSKIDVNGDGELSLEEFIEGVQKDQMLLDTLTRSLDLTRIVRRLQNGEQDEEGADEAAEAAG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf131 chromosome 6 open reading frame 131 CORD14 GCAP 1 GCAP1photoreceptor 1 GCAPGUCA Guanylate cyclase activator 1A guanylate cyclase activator 1A (retina) guanylin 1 retina guanylyl cyclase-activating protein 1 GUCA1; cone dystrophy 3; GCAP 1; Guanylate cyclase activator 1A; guanylate cyclase-activating protein, photoreceptor 1; guanylin 1, retina; Guanylyl cyclase-activating protein 1
Gene Aliases: C6orf131; COD3; CORD14; GCAP; GCAP1; GUCA; GUCA1; GUCA1A
UniProt ID: (Human) P43080
Entrez Gene ID: (Human) 2978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.