Proteintech
Manufacturer Code:129441AP150UL
Catalog # 50-172-8366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GCAP1 Fusion Protein Ag4017 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf131 COD3 GCAP GCAP 1 GCAP1 Guanylate cyclase activator 1A GUCA GUCA1 GUCA1A; cone dystrophy 3; GCAP 1; Guanylate cyclase activator 1A; guanylate cyclase-activating protein, photoreceptor 1; guanylin 1, retina; Guanylyl cyclase-activating protein 1
Gene Aliases: C6orf131; COD3; CORD14; GC-A; GCAP; GCAP1; GUCA; GUCA1; GUCA1A; mGCAP1
UniProt ID: (Human) P43080, (Mouse) P43081
Entrez Gene ID: (Human) 2978, (Mouse) 14913, (Rat) 301233
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.