Invitrogen
Catalog # HYB 249-05-02
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | 1:2,000 |
Host / Isotype | Mouse / IgG2b |
Class | Monoclonal |
Type | Antibody |
Clone | 19E8 |
Immunogen | Gc-globulin isolated from human plasma. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | 0.93 mg/mL |
Storage conditions | 4° C, store in dark |
RRID | AB_1074572 |
HYB 249-05-02 detects Gc-Globulin in human samples.
HYB 249-05-02 has been successfully used in ELISA techniques. HYB 249-05-02 reacts strongly with Gc-globulin when tested in sandwich ELISA in combination with a polyclonal antibody against Gc-globulin.
The HYB 249-05-02 immunogen is Gc-Globulin found in human plasma cells.
Epitope differs from that of HYB 249-01-02, HYB 249-02-02 and HYB 249-10-02.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.